accessibility menu, dialog, popup

UserName

H3COBr

CAS RN 174702-59-1

IUPAC Name: 4-bromo-2-methyl-2,3-dihydro-1H-inden-1-one
Synonyms: 4-bromo-2-methyl-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 225.09
Molecular Formula: C10H9BrO
InChi Key: AHMJTIYXKJRVFF-UHFFFAOYNA-N
SMILES: CC1CC2=C(C=CC=C2Br)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results