missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methyl-1-indanone 98.0+%, TCI America™

Catalog No. B50965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50965G 5 g
B50961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50965G Supplier TCI America Supplier No. B50965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 174702-59-1
Molecular Formula C10H9BrO
Molecular Weight (g/mol) 225.085
MDL Number MFCD09881709
InChI Key AHMJTIYXKJRVFF-UHFFFAOYSA-N
PubChem CID 10537152
IUPAC Name 4-bromo-2-methyl-2,3-dihydroinden-1-one
SMILES CC1CC2=C(C1=O)C=CC=C2Br

Specifications

Color White-Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 225.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-2-methyl-1-indanone