missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-hydroxybenzonitrile 98.0+%, TCI America™
$42.36 - $167.49
Chemical Identifiers
| CAS | 288067-35-6 |
|---|---|
| Molecular Formula | C7H4BrNO |
| Molecular Weight (g/mol) | 198.019 |
| MDL Number | MFCD07780675 |
| InChI Key | PAHSHGVACWNGEY-UHFFFAOYSA-N |
| Synonym | 5-Bromo-2-cyanophenol |
| PubChem CID | 12966961 |
| IUPAC Name | 4-bromo-2-hydroxybenzonitrile |
| SMILES | C1=CC(=C(C=C1Br)O)C#N |
Chemical Identifiers
| 288067-35-6 | |
| 198.019 | |
| PAHSHGVACWNGEY-UHFFFAOYSA-N | |
| 12966961 | |
| C1=CC(=C(C=C1Br)O)C#N |
| C7H4BrNO | |
| MFCD07780675 | |
| 5-Bromo-2-cyanophenol | |
| 4-bromo-2-hydroxybenzonitrile |
Specifications
| 288067-35-6 | |
| C7H4BrNO | |
| 1 g | |
| 5-Bromo-2-cyanophenol | |
| C1=CC(=C(C=C1Br)O)C#N | |
| 198.019 | |
| 198.02 | |
| Crystalline Powder |
| 160°C | |
| MFCD07780675 | |
| 3439 | |
| PAHSHGVACWNGEY-UHFFFAOYSA-N | |
| 4-bromo-2-hydroxybenzonitrile | |
| 12966961 | |
| ≥98.0% (GC) | |
| 4-Bromo-2-hydroxybenzonitrile |
Safety and Handling
TSCA : No