missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromo-2'-hydroxyacetophenone 98.0+%, TCI America™
$41.02 - $121.16
Chemical Identifiers
| CAS | 30186-18-6 |
|---|---|
| Molecular Formula | C8H7BrO2 |
| Molecular Weight (g/mol) | 215.05 |
| MDL Number | MFCD03428533 |
| InChI Key | LQCMMXGKEGWUIM-UHFFFAOYSA-N |
| Synonym | 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone |
| PubChem CID | 10727403 |
| IUPAC Name | 1-(4-bromo-2-hydroxyphenyl)ethan-1-one |
| SMILES | CC(=O)C1=C(O)C=C(Br)C=C1 |
Chemical Identifiers
| 30186-18-6 | |
| 215.05 | |
| LQCMMXGKEGWUIM-UHFFFAOYSA-N | |
| 10727403 | |
| CC(=O)C1=C(O)C=C(Br)C=C1 |
| C8H7BrO2 | |
| MFCD03428533 | |
| 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone | |
| 1-(4-bromo-2-hydroxyphenyl)ethan-1-one |
Specifications
| 30186-18-6 | |
| 83°C | |
| MFCD03428533 | |
| 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone | |
| CC(=O)C1=C(O)C=C(Br)C=C1 | |
| 215.05 | |
| 215.05 | |
| Crystalline Powder |
| 42°C | |
| C8H7BrO2 | |
| 1 g | |
| LQCMMXGKEGWUIM-UHFFFAOYSA-N | |
| 1-(4-bromo-2-hydroxyphenyl)ethan-1-one | |
| 10727403 | |
| ≥98.0% (GC,T) | |
| 4′-Bromo-2′-hydroxyacetophenone |
Safety and Handling
TSCA : No