missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-hydroxyacetophenone, 95%

CAS: 30186-18-6 | C8H7BrO2 | 215.05 g/mol

$158.71 - $616.10

Chemical Identifiers

CAS 30186-18-6
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD03428533
InChI Key LQCMMXGKEGWUIM-UHFFFAOYSA-N
Synonym 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone
PubChem CID 10727403
IUPAC Name 1-(4-bromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6113703
View Documents
Thermo Scientific Chemicals
H6113703
1 g
Each for $158.71
Only null left
 
AAH6113706
View Documents
Thermo Scientific Chemicals
H6113706
5 g
Each for $616.10
Only null left
 
Description

Description

4'-Bromo-2'-hydroxyacetophenone is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromo-2′-hydroxyacetophenone is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

30186-18-6
215.05
LQCMMXGKEGWUIM-UHFFFAOYSA-N
10727403
CC(=O)C1=C(O)C=C(Br)C=C1
C8H7BrO2
MFCD03428533
1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone
1-(4-bromo-2-hydroxyphenyl)ethanone

Specifications

30186-18-6
MFCD03428533
1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone
LQCMMXGKEGWUIM-UHFFFAOYSA-N
1-(4-bromo-2-hydroxyphenyl)ethanone
10727403
95%
4'-Bromo-2'-hydroxyacetophenone
C8H7BrO2
1 g
Slightly soluble in water.
CC(=O)C1=C(O)C=C(Br)C=C1
215.05
215.04
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only