missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-furaldehyde 97.0+%, TCI America™
$452.97 - $1543.44
Chemical Identifiers
| CAS | 21921-76-6 |
|---|---|
| Molecular Formula | C5H3BrO2 |
| Molecular Weight (g/mol) | 174.981 |
| MDL Number | MFCD00671504 |
| InChI Key | MRGBBKQOSUHKPF-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-furaldehyde, 4-bromofurfural, 4-bromo-2-formylfuran, 4-bromo-2-furancarbaldehyde, 2-furancarboxaldehyde, 4-bromo, 4-bromo-furfural, 4-bromofuraldehyde, zlchem 82, pubchem3545, acmc-209fpi |
| PubChem CID | 2757010 |
| IUPAC Name | 4-bromofuran-2-carbaldehyde |
| SMILES | C1=C(OC=C1Br)C=O |
Chemical Identifiers
| 21921-76-6 | |
| 174.981 | |
| MRGBBKQOSUHKPF-UHFFFAOYSA-N | |
| 2757010 | |
| C1=C(OC=C1Br)C=O |
| C5H3BrO2 | |
| MFCD00671504 | |
| 4-bromo-2-furaldehyde, 4-bromofurfural, 4-bromo-2-formylfuran, 4-bromo-2-furancarbaldehyde, 2-furancarboxaldehyde, 4-bromo, 4-bromo-furfural, 4-bromofuraldehyde, zlchem 82, pubchem3545, acmc-209fpi | |
| 4-bromofuran-2-carbaldehyde |
Specifications
| 21921-76-6 | |
| White-Yellow | |
| MFCD00671504 | |
| 4-bromo-2-furaldehyde, 4-bromofurfural, 4-bromo-2-formylfuran, 4-bromo-2-furancarbaldehyde, 2-furancarboxaldehyde, 4-bromo, 4-bromo-furfural, 4-bromofuraldehyde, zlchem 82, pubchem3545, acmc-209fpi | |
| C1=C(OC=C1Br)C=O | |
| 174.981 | |
| 174.98 | |
| Crystalline Powder |
| 55°C | |
| C5H3BrO2 | |
| 1 g | |
| MRGBBKQOSUHKPF-UHFFFAOYSA-N | |
| 4-bromofuran-2-carbaldehyde | |
| 2757010 | |
| ≥97.0% (GC) | |
| 4-Bromo-2-furaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator