missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-2-furaldehyde 97.0+%, TCI America™

Manufacturer:  TCI America B18165G

 View more versions of this product

Catalog No. B18165G



Specifications

Specifications

4-Bromo-2-furaldehyde
55°C
21921-76-6
MFCD00671504
MRGBBKQOSUHKPF-UHFFFAOYSA-N
4-bromofuran-2-carbaldehyde
2757010
Crystal-Powder at 20°C
White-Yellow
5g
C5H3BrO2
4-bromo-2-furaldehyde, 4-bromofurfural, 4-bromo-2-formylfuran, 4-bromo-2-furancarbaldehyde, 2-furancarboxaldehyde, 4-bromo, 4-bromo-furfural, 4-bromofuraldehyde, zlchem 82, pubchem3545, acmc-209fpi
C1=C(OC=C1Br)C=O
174.981
174.98
≥97.0% (GC)

Chemical Identifiers

21921-76-6
174.981
MRGBBKQOSUHKPF-UHFFFAOYSA-N
2757010
C1=C(OC=C1Br)C=O
C5H3BrO2
MFCD00671504
4-bromo-2-furaldehyde, 4-bromofurfural, 4-bromo-2-formylfuran, 4-bromo-2-furancarbaldehyde, 2-furancarboxaldehyde, 4-bromo, 4-bromo-furfural, 4-bromofuraldehyde, zlchem 82, pubchem3545, acmc-209fpi
4-bromofuran-2-carbaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents