missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoropyridine 98.0+%, TCI America™

Catalog No. B47781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47781G 1 g
B47785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47781G Supplier TCI America Supplier No. B47781G
Only null left

Chemical Identifiers

CAS 128071-98-7
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD04112504
InChI Key PTPTZLXZHPPVKG-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromopyridine, 4-bromo-2-fluoro-pyridine, pyridine, 4-bromo-2-fluoro, abbypharma ap-17-5099, pubchem1228, acmc-209bdf, pyff21, 4-bromo-2-fluoro pyridine, ksc493i4r, 4-bromanyl-2-fluoranyl-pyridine
PubChem CID 2762851
IUPAC Name 4-bromo-2-fluoropyridine
SMILES C1=CN=C(C=C1Br)F

Specifications

Color Yellow
Boiling Point 65°C
Quantity 1 g
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluoropyridine