missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobiphenyl 99.0+%, TCI America™

Catalog No. B20285G
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20285G 5 g
B202825G 25 g
B2028250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B20285G Supplier TCI America Supplier No. B20285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41604-19-7
Molecular Formula C12H8BrF
Molecular Weight (g/mol) 251.10
MDL Number MFCD00051716
InChI Key HTRNHWBOBYFTQF-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
PubChem CID 521063
IUPAC Name 4-bromo-2-fluoro-1,1'-biphenyl
SMILES FC1=CC(Br)=CC=C1C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3152
Formula Weight 251.10
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-2-fluorobiphenyl