missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzyl cyanide, 98+%, Thermo Scientific™

Catalog No. p-3212014
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC384450050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC384450050 Supplier Thermo Scientific Chemicals Supplier No. 384450050

Chemical Identifiers

CAS 114897-91-5
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.04
MDL Number MFCD07368653
InChI Key QLASQEZPJFNZQC-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzyl cyanide, 2-4-bromo-2-fluorophenyl acetonitrile, 4-bromo-2-fluorophenylacetonitrile, 2-fluoro-4-bromophenylacetonitrile, benzeneacetonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorophenyl acetonitrile, 2-4-bromo-2-fluorophenyl ethanenitrile, 4-bromo-2-fluorobenzylcyanide, pubchem10413, acmc-2099lv
PubChem CID 7172308
IUPAC Name 2-(4-bromo-2-fluorophenyl)acetonitrile
SMILES FC1=CC(Br)=CC=C1CC#N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 52.8°C to 53.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CNCH2C6H3(F)Br
Quantity 5 g
Formula Weight 214.04
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluorobenzyl cyanide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only