missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzyl bromide, 98%

Catalog No. p-7025696
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2133106 5 g
AAB2133114 25 g
AAB2133122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2133106 Supplier Thermo Scientific Chemicals Supplier No. B2133106

CAS: 76283-09-5 | C7H5Br2F | 267.923 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76283-09-5
Molecular Formula C7H5Br2F
Molecular Weight (g/mol) 267.923
MDL Number MFCD00055467
InChI Key XMHNLZXYPAULDF-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzyl bromide, 4-bromo-1-bromomethyl-2-fluorobenzene, 2-fluoro-4-bromobenzyl bromide, 4-bromo-2-fluorobenzylbromide, 2-fluoro-4-bromo-benzylbromide, alpha,4-dibromo-2-fluorotoluene, unii-0z26xyc1jy, 4,alpha-dibromo-2-fluorotoluene, 4-bromo-1-bromomethyl-2-fluoro-benzene, 0z26xyc1jy
PubChem CID 2733660
IUPAC Name 4-bromo-1-(bromomethyl)-2-fluorobenzene
SMILES C1=CC(=C(C=C1Br)F)CBr

Specifications

Melting Point 33°C to 37°C
Boiling Point 126°C (9 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 4307676
Formula Weight 267.92
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzyl bromide
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 278-412-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only