missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog No. AAL1867906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1867906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1867906 Supplier Thermo Scientific Chemicals Supplier No. L1867906
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 188582-62-9
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD00143265
InChI Key BWBJZMQPVBWEJU-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzyl alcohol, 4-bromo-2-fluorophenyl methanol, 4-bromo-2-fluorobenzylalcohol, benzenemethanol, 4-bromo-2-fluoro, rarechem al bd 0386, timtec-bb sbb005826, 4-bromo-2-fluorophenyl methan-1-ol, 2-fluoro-4-bromobenzyl alcohol, pubchem3223, acmc-209erg
PubChem CID 2734027
IUPAC Name (4-bromo-2-fluorophenyl)methanol
SMILES C1=CC(=C(C=C1Br)F)CO

Specifications

Melting Point 31°C to 33°C
Quantity 5 g
Beilstein 7757667
Formula Weight 205.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluorobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only