missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. B29465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29465G 5 g
B294625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29465G Supplier TCI America Supplier No. B29465G

Chemical Identifiers

CAS 142808-15-9
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD01075254
InChI Key OEPBVXQEVBURGC-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzotrifluoride, 4-bromo-2-fluoro-1-trifluoromethyl benzene, 2-fluoro-4-bromobenzotrifluoride, 1-bromo-3-fluoro-4-trifluoromethyl benzene, benzene, 4-bromo-2-fluoro-1-trifluoromethyl, 2-fluoro-4-bromotrifluorotoluene, 3-fluoro-4-trifluoromethyl bromobenzene, 4-bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 4-bromo-2-fluoro benzotrifluoride, 4-bromo-2-fluoro-1-trifluoromethyl-benzene
PubChem CID 2782904
IUPAC Name 4-bromo-2-fluoro-1-(trifluoromethyl)benzene
SMILES FC1=C(C=CC(Br)=C1)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 243.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluorobenzotrifluoride