missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7119747
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27455G 5 g
B274525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27455G Supplier TCI America Supplier No. B27455G
Only null left

Chemical Identifiers

CAS 105942-08-3
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00143264
InChI Key HGXWRDPQFZKOLZ-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzonitrile, 4-bromo-2-fluoro-benzonitrile, benzonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorobenzenecarbonitrile, 4-cyano-3-fluorobromobenzene, pubchem3221, 2-floro-4-bromobenzonitrile, 4-bromo-2-fluorobenzontrile, 4-bromo-6-fluorobenzonitrile, ksc490q4t
PubChem CID 736029
IUPAC Name 4-bromo-2-fluorobenzonitrile
SMILES FC1=CC(Br)=CC=C1C#N

Specifications

Melting Point 71°C
Boiling Point 239°C
Quantity 5 g
UN Number 3439
Formula Weight 200.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluorobenzonitrile