missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzonitrile, 99%

Catalog No. p-7020954
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1308606 5 g
AAA1308614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1308606 Supplier Thermo Scientific Chemicals Supplier No. A1308606
Only null left

CAS: 105942-08-3 | C7H3BrFN | 200.01 g/mol

4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 105942-08-3
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00143264
InChI Key HGXWRDPQFZKOLZ-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzonitrile, 4-bromo-2-fluoro-benzonitrile, benzonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorobenzenecarbonitrile, 4-cyano-3-fluorobromobenzene, pubchem3221, 2-floro-4-bromobenzonitrile, 4-bromo-2-fluorobenzontrile, 4-bromo-6-fluorobenzonitrile, ksc490q4t
PubChem CID 736029
IUPAC Name 4-bromo-2-fluorobenzonitrile
SMILES FC1=CC(Br)=CC=C1C#N

Specifications

Melting Point 69°C to 72°C
Boiling Point 238°C to 239°C
Quantity 5 g
UN Number UN3439
Beilstein 4231143
Solubility Information Insoluble in water.
Formula Weight 200.01
Percent Purity 99%
Chemical Name or Material 4-Bromo-2-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only