missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-fluorobenzoic Acid 99.0+%, TCI America™
$59.69 - $221.39
Chemical Identifiers
| CAS | 112704-79-7 |
|---|---|
| Molecular Formula | C7H3BrFO2 |
| Molecular Weight (g/mol) | 218.00 |
| MDL Number | MFCD00143263 |
| InChI Key | ZQQSRVPOAHYHEL-UHFFFAOYSA-M |
| Synonym | 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754 |
| PubChem CID | 302681 |
| IUPAC Name | 4-bromo-2-fluorobenzoate |
| SMILES | [O-]C(=O)C1=CC=C(Br)C=C1F |
Chemical Identifiers
| 112704-79-7 | |
| 218.00 | |
| ZQQSRVPOAHYHEL-UHFFFAOYSA-M | |
| 302681 | |
| [O-]C(=O)C1=CC=C(Br)C=C1F |
| C7H3BrFO2 | |
| MFCD00143263 | |
| 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754 | |
| 4-bromo-2-fluorobenzoate |
Specifications
| 112704-79-7 | |
| White-Yellow | |
| MFCD00143263 | |
| 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754 | |
| [O-]C(=O)C1=CC=C(Br)C=C1F | |
| 218.00 | |
| 219.01 | |
| Crystalline Powder |
| 212°C | |
| C7H3BrFO2 | |
| 5 g | |
| ZQQSRVPOAHYHEL-UHFFFAOYSA-M | |
| 4-bromo-2-fluorobenzoate | |
| 302681 | |
| ≥99.0% (T) | |
| 4-Bromo-2-fluorobenzoic Acid |
Safety and Handling
TSCA : No