missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzoic Acid 99.0+%, TCI America™

Catalog No. B19505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19505G 5 g
B195025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19505G Supplier TCI America Supplier No. B19505G

Chemical Identifiers

CAS 112704-79-7
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00143263
InChI Key ZQQSRVPOAHYHEL-UHFFFAOYSA-M
Synonym 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754
PubChem CID 302681
IUPAC Name 4-bromo-2-fluorobenzoate
SMILES [O-]C(=O)C1=CC=C(Br)C=C1F

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.01
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluorobenzoic Acid