missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-fluorobenzaldehyde 96.0+%, TCI America™
$132.84 - $377.33
Chemical Identifiers
| CAS | 57848-46-1 |
|---|---|
| Molecular Formula | C7H4BrFO |
| Molecular Weight (g/mol) | 203.01 |
| MDL Number | MFCD00143261 |
| InChI Key | UPCARQPLANFGQJ-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde |
| PubChem CID | 143517 |
| IUPAC Name | 4-bromo-2-fluorobenzaldehyde |
| SMILES | FC1=CC(Br)=CC=C1C=O |
Chemical Identifiers
| 57848-46-1 | |
| 203.01 | |
| UPCARQPLANFGQJ-UHFFFAOYSA-N | |
| 143517 | |
| FC1=CC(Br)=CC=C1C=O |
| C7H4BrFO | |
| MFCD00143261 | |
| 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde | |
| 4-bromo-2-fluorobenzaldehyde |
Specifications
| 57848-46-1 | |
| White-Yellow | |
| MFCD00143261 | |
| 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde | |
| FC1=CC(Br)=CC=C1C=O | |
| 203.01 | |
| 203.01 | |
| Crystalline Powder |
| 63°C | |
| C7H4BrFO | |
| 5 g | |
| UPCARQPLANFGQJ-UHFFFAOYSA-N | |
| 4-bromo-2-fluorobenzaldehyde | |
| 143517 | |
| ≥96.0% (GC) | |
| 4-Bromo-2-fluorobenzaldehyde |
Safety and Handling
TSCA : No