missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzaldehyde 96.0+%, TCI America™

Catalog No. B20025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20025G 5 g
B200225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20025G Supplier TCI America Supplier No. B20025G

Chemical Identifiers

CAS 57848-46-1
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00143261
InChI Key UPCARQPLANFGQJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde
PubChem CID 143517
IUPAC Name 4-bromo-2-fluorobenzaldehyde
SMILES FC1=CC(Br)=CC=C1C=O

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 203.01
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-fluorobenzaldehyde