Learn More
4-Bromo-2-fluorobenzaldehyde, 97%
CAS: 57848-46-1 | C7H4BrFO | 203.01 g/mol
$84.01 - $300.10
Chemical Identifiers
| CAS | 57848-46-1 |
|---|---|
| Molecular Formula | C7H4BrFO |
| Molecular Weight (g/mol) | 203.01 |
| MDL Number | MFCD00143261 |
| InChI Key | UPCARQPLANFGQJ-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde |
| PubChem CID | 143517 |
| IUPAC Name | 4-bromo-2-fluorobenzaldehyde |
| SMILES | FC1=CC(Br)=CC=C1C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1611206
|
Thermo Scientific Chemicals
A1611206 |
5 g |
Each for $84.01
|
|
|||||
|
AAA1611214
|
Thermo Scientific Chemicals
A1611214 |
25 g |
Each for $300.10
|
|
|||||
Description
4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.
Solubility
Insoluble in water.
Notes
Air and light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases. Keep the container tightly closed.
Chemical Identifiers
| 57848-46-1 | |
| 203.01 | |
| UPCARQPLANFGQJ-UHFFFAOYSA-N | |
| 143517 | |
| FC1=CC(Br)=CC=C1C=O |
| C7H4BrFO | |
| MFCD00143261 | |
| 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde | |
| 4-bromo-2-fluorobenzaldehyde |
Specifications
| 57848-46-1 | |
| 42°C (19 mmHg) | |
| MFCD00143261 | |
| 7700208 | |
| 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde | |
| UPCARQPLANFGQJ-UHFFFAOYSA-N | |
| 4-bromo-2-fluorobenzaldehyde | |
| 143517 | |
| 97% |
| 59°C to 63°C | |
| C7H4BrFO | |
| 5 g | |
| Air Sensitive | |
| Insoluble in water. | |
| FC1=CC(Br)=CC=C1C=O | |
| 203.01 | |
| 203.01 | |
| 4-Bromo-2-fluorobenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only