missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzaldehyde, 97%

Catalog No. AAA1611206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1611206 5 g
AAA1611214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1611206 Supplier Thermo Scientific Chemicals Supplier No. A1611206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57848-46-1 | C7H4BrFO | 203.01 g/mol

4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzaldehyde is used in the preparation of nigeglanine hydrobromide.

Solubility
Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases. Keep the container tightly closed.

Chemical Identifiers

CAS 57848-46-1
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00143261
InChI Key UPCARQPLANFGQJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2-fluoro, 4-bromo-2-fluoro-benzaldehyde, pubchem1421, acmc-1awi3, 4-bromo-2-flurobenzaldehyde, 4-bromo-2 fluorobenzaldehyde, ksc272o0t, 2-fluoro-4-bromo-benzaldehyde, 4-bromo-2-fluoro benzaldehyde
PubChem CID 143517
IUPAC Name 4-bromo-2-fluorobenzaldehyde
SMILES FC1=CC(Br)=CC=C1C=O

Specifications

Melting Point 59°C to 63°C
Boiling Point 42°C (19 mmHg)
Quantity 5 g
Beilstein 7700208
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 203.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only