missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoro-6-nitroanisole, 97%

Catalog No. AAH3022006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3022006 5 g
AAH3022014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3022006 Supplier Thermo Scientific Chemicals Supplier No. H3022006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 74266-66-3 | C7H5BrFNO3 | 250.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74266-66-3
Molecular Formula C7H5BrFNO3
Molecular Weight (g/mol) 250.02
MDL Number MFCD00013374
InChI Key CVCAYLYLJKBSNV-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoro-6-nitroanisole, 5-bromo-3-fluoro-2-methoxy-1-nitrobenzene, 5-bromo-1-fluoro-2-methoxy-3-nitro-benzene, benzene, 5-bromo-1-fluoro-2-methoxy-3-nitro, 4-bromo-2-fluoro-6-nitrophenyl methyl ether, timtec-bb sbb005948, acmc-20amnn, pubchem4134, 4-bromo-2-fluoro-6-nitroanisol
PubChem CID 557159
IUPAC Name 5-bromo-1-fluoro-2-methoxy-3-nitrobenzene
SMILES COC1=C(F)C=C(Br)C=C1[N+]([O-])=O

Specifications

Melting Point 53°C to 56°C
Quantity 5 g
Formula Weight 250.02
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluoro-6-nitroanisole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only