missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-ethyl-1-iodobenzene, 97%, stab. with copper, Thermo Scientific Chemicals

Catalog No. p-7024824
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2005288 2.5 g
AAB2005209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAB2005288 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' B2005288
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175278-30-5
Molecular Formula C8H8BrI
Molecular Weight (g/mol) 310.96
MDL Number MFCD00221458
InChI Key JWKQXPHYKRQLEJ-UHFFFAOYSA-N
PubChem CID 2761396
IUPAC Name 4-bromo-2-ethyl-1-iodobenzene
SMILES CCC1=C(C=CC(=C1)Br)I

Specifications

Boiling Point 84°C to 86°C (0.4 mmHg)
Quantity 2.5 g
Sensitivity Light sensitive
Synonym 4-bromo-2-ethyliodobenzene, benzene,4-bromo-2-ethyl-1-iodo, benzene, 4-bromo-2-ethyl-1-iodo, 4-bromo-2-ethyl-1-iodo-benzene, pubchem3773, acmc-1c9j2, 4-bromo-2-ethyl-iodobenzene, 1-iodo-2-ethyl-4-bromobenzene, 4-bromo-2-ethyl-1-iodobenzene, 4-bromanyl-2-ethyl-1-iodanyl-benzene
Formula Weight 310.97
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-ethyl-1-iodobenzene, stabilized with copper
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only