missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-chlorotoluene 97.0+%, TCI America™
$44.73 - $108.11
Chemical Identifiers
| CAS | 89794-02-5 |
|---|---|
| Molecular Formula | C7H6BrCl |
| Molecular Weight (g/mol) | 205.479 |
| MDL Number | MFCD00060651 |
| InChI Key | LIFMTDJMLRECMX-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-chlorotoluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, benzene, 4-bromo-2-chloro-1-methyl, 4-bromo-chlorotoluene, pubchem3603, 4-bromo-2chlorotoluene, 2-chloro 4-bromo toluene, acmc-209r2x |
| PubChem CID | 2735554 |
| IUPAC Name | 4-bromo-2-chloro-1-methylbenzene |
| SMILES | CC1=C(C=C(C=C1)Br)Cl |
Chemical Identifiers
| 89794-02-5 | |
| 205.479 | |
| LIFMTDJMLRECMX-UHFFFAOYSA-N | |
| 2735554 | |
| CC1=C(C=C(C=C1)Br)Cl |
| C7H6BrCl | |
| MFCD00060651 | |
| 4-bromo-2-chlorotoluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, benzene, 4-bromo-2-chloro-1-methyl, 4-bromo-chlorotoluene, pubchem3603, 4-bromo-2chlorotoluene, 2-chloro 4-bromo toluene, acmc-209r2x | |
| 4-bromo-2-chloro-1-methylbenzene |
Specifications
| 89794-02-5 | |
| 5 g | |
| C7H6BrCl | |
| 4-bromo-2-chlorotoluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, benzene, 4-bromo-2-chloro-1-methyl, 4-bromo-chlorotoluene, pubchem3603, 4-bromo-2chlorotoluene, 2-chloro 4-bromo toluene, acmc-209r2x | |
| CC1=C(C=C(C=C1)Br)Cl | |
| 205.479 | |
| 205.48 | |
| 4-Bromo-2-chlorotoluene |
| 110°C | |
| Clear Liquid at 20°C | |
| MFCD00060651 | |
| LIFMTDJMLRECMX-UHFFFAOYSA-N | |
| 4-bromo-2-chloro-1-methylbenzene | |
| 2735554 | |
| ≥97.0% (GC) |
Safety and Handling
TSCA : No