missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzonitrile, 96%

Catalog No. p-7058347
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1060203 1 g
AAL1060206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1060203 Supplier Thermo Scientific Chemicals Supplier No. L1060203
Only null left

CAS: 154607-01-9 | C7H3BrClN | 216.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154607-01-9
Molecular Formula C7H3BrClN
Molecular Weight (g/mol) 216.46
MDL Number MFCD00040883
InChI Key AYQBMZNSJPVADT-UHFFFAOYSA-N
Synonym 2-chloro-4-bromobenzonitrile, 4-bromo-2-chloro-benzonitrile, benzonitrile, 4-bromo-2-chloro, 4-bromo-2-chlorobenzenecarbonitrile, pubchem12801, intermediates-zcf02177, acmc-209db5, ksc497g8d
PubChem CID 7010420
IUPAC Name 4-bromo-2-chlorobenzonitrile
SMILES ClC1=C(C=CC(Br)=C1)C#N

Specifications

Melting Point 67°C to 72°C
Boiling Point 142°C to 143°C (10 mmHg)
Quantity 1 g
UN Number UN3439
Beilstein 3240885
Formula Weight 216.47
Percent Purity 96%
Chemical Name or Material 4-Bromo-2-chlorobenzonitrile
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only