missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7119037
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14071G 1 g
B14075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14071G Supplier TCI America Supplier No. B14071G

Chemical Identifiers

CAS 59748-90-2
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00040903
InChI Key JAVZWSOFJKYSDY-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d
PubChem CID 108829
IUPAC Name 4-bromo-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Br)Cl)C(=O)O

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 1 g
Formula Weight 235.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-chlorobenzoic Acid