missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-chlorobenzaldehyde 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 75°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Synonym | 2-chloro-4-bromobenzaldehyde, 4-bromo-2-chloro-benzaldehyde, benzaldehyde, 4-bromo-2-chloro, acmc-1c7dt, intermediates-zcf02199, ksc497g8p, benzaldehyde,4-bromo-2-chloro, 4-bromanyl-2-chloranyl-benzaldehyde, 2-chloro-4-bromobenzaldehyde, 95+% |
| Formula Weight | 219.46 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromo-2-chlorobenzaldehyde |
Safety and Handling
| TSCA | No |