missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-6-methylaniline, 98%

Catalog No. AAL1259006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1259006 5 g
AAL1259014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1259006 Supplier Thermo Scientific Chemicals Supplier No. L1259006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30273-42-8 | C7H7BrClN | 220.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30273-42-8
Molecular Formula C7H7BrClN
Molecular Weight (g/mol) 220.49
MDL Number MFCD00041432
InChI Key DIXGIKZIIZRFKE-UHFFFAOYSA-N
Synonym 4-bromo-2-chloro-6-methyl-phenylamine, benzenamine, 4-bromo-2-chloro-6-methyl, pubchem23742, acmc-20a53i, 4-bromo-6-chloro-o-toluidine, 2-amino-5-bromo-3-chlorotoluene, 4-bromo-2-chloro-6-methyl-aniline, 4-bromo-6-chloro-2-methylphenylamine, 4-bromo-2-chloro-6-methylphenylamine, benzenamine,4-bromo-2-chloro-6-methyl
PubChem CID 2769626
IUPAC Name 4-bromo-2-chloro-6-methylaniline
SMILES CC1=CC(Br)=CC(Cl)=C1N

Specifications

Melting Point 37°C to 40°C
Quantity 5 g
UN Number UN2811
Beilstein 2717133
Formula Weight 220.5
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chloro-6-methylaniline
Hazard Category H302+H312+H332-H373
Hazard Statement GHS H Statement
H331-H373-H302-H312
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P314-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only