missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,6-diphenylpyrimidine 98.0+%, TCI America™
$96.26 - $323.43
Chemical Identifiers
| CAS | 40734-24-5 |
|---|---|
| Molecular Formula | C16H11BrN2 |
| Molecular Weight (g/mol) | 311.182 |
| MDL Number | MFCD00234885 |
| InChI Key | IJXDLFJKGQNBEJ-UHFFFAOYSA-N |
| PubChem CID | 817533 |
| IUPAC Name | 4-bromo-2,6-diphenylpyrimidine |
| SMILES | C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4943200MG
|
TCI America
B4943200MG |
200 mg |
Each for $96.26
|
|
|||||
|
B49431G
|
TCI America
B49431G |
1 g |
Each for $323.43
|
|
|||||
Chemical Identifiers
| 40734-24-5 | |
| 311.182 | |
| IJXDLFJKGQNBEJ-UHFFFAOYSA-N | |
| 4-bromo-2,6-diphenylpyrimidine |
| C16H11BrN2 | |
| MFCD00234885 | |
| 817533 | |
| C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)Br |
Specifications
| 40734-24-5 | |
| White | |
| MFCD00234885 | |
| IJXDLFJKGQNBEJ-UHFFFAOYSA-N | |
| 4-bromo-2,6-diphenylpyrimidine | |
| 817533 | |
| ≥98.0% (GC) | |
| 4-Bromo-2,6-diphenylpyrimidine |
| 112°C | |
| C16H11BrN2 | |
| 200 mg | |
| C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)Br | |
| 311.182 | |
| 311.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No