accessibility menu, dialog, popup

UserName

NNBr

CAS RN 40734-24-5

IUPAC Name: 4-bromo-2,6-diphenylpyrimidine
Synonyms: 4-bromo-2,6-diphenylpyrimidine
Molecular Weight (g/mol): 311.18
Molecular Formula: C16H11BrN2
InChi Key: IJXDLFJKGQNBEJ-UHFFFAOYSA-N
SMILES: BrC1=NC(=NC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results