missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,6-diphenylpyrimidine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 112°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 311.18 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromo-2,6-diphenylpyrimidine |
Safety and Handling
| TSCA | No |