missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorophenylacetic acid, 96%, Thermo Scientific™

$347.77 - $347.77

Chemical Identifiers

CAS 537033-54-8
Molecular Formula C8H5BrF2O2
Molecular Weight (g/mol) 251.03
MDL Number MFCD03094466
InChI Key GBFFYYLXGIHKLU-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluorophenylacetic acid, 2-4-bromo-2,6-difluorophenyl acetic acid, 4-bromo-2,6-difluorophenyl acetic acid, 2-4-bromo-2,6-difluoro-phenyl acetic acid, acmc-209uz8, 4-bromo-2,6-difluorophenylaceticacid, 4-bromo-2,6-difluorobenzeneacetic acid, 4-bromo-2,6-difluoro-phenyl-acetic acid, benzeneacetic acid, 4-bromo-2,6-difluoro
PubChem CID 2773309
IUPAC Name 2-(4-bromo-2,6-difluorophenyl)acetic acid
SMILES OC(=O)CC1=C(F)C=C(Br)C=C1F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6623103
View Documents
Thermo Scientific Chemicals
H6623103
1 g
Each for $347.77
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

537033-54-8
251.03
GBFFYYLXGIHKLU-UHFFFAOYSA-N
2773309
OC(=O)CC1=C(F)C=C(Br)C=C1F
C8H5BrF2O2
MFCD03094466
4-bromo-2,6-difluorophenylacetic acid, 2-4-bromo-2,6-difluorophenyl acetic acid, 4-bromo-2,6-difluorophenyl acetic acid, 2-4-bromo-2,6-difluoro-phenyl acetic acid, acmc-209uz8, 4-bromo-2,6-difluorophenylaceticacid, 4-bromo-2,6-difluorobenzeneacetic acid, 4-bromo-2,6-difluoro-phenyl-acetic acid, benzeneacetic acid, 4-bromo-2,6-difluoro
2-(4-bromo-2,6-difluorophenyl)acetic acid

Specifications

537033-54-8
C8H5BrF2O2
1 g
GBFFYYLXGIHKLU-UHFFFAOYSA-N
2-(4-bromo-2,6-difluorophenyl)acetic acid
2773309
96%
4-Bromo-2,6-difluorophenylacetic acid
1.77 g/mL
MFCD03094466
4-bromo-2,6-difluorophenylacetic acid, 2-4-bromo-2,6-difluorophenyl acetic acid, 4-bromo-2,6-difluorophenyl acetic acid, 2-4-bromo-2,6-difluoro-phenyl acetic acid, acmc-209uz8, 4-bromo-2,6-difluorophenylaceticacid, 4-bromo-2,6-difluorobenzeneacetic acid, 4-bromo-2,6-difluoro-phenyl-acetic acid, benzeneacetic acid, 4-bromo-2,6-difluoro
OC(=O)CC1=C(F)C=C(Br)C=C1F
251.03
251.02
Solid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only