missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dichlorobenzenesulfonyl chloride, 95%

Catalog No. aal1987906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1987906 5 g
AAL1987903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1987906 Supplier Thermo Scientific Chemicals Supplier No. L1987906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 351003-54-8 | C6H2BrCl3O2S | 324.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 351003-54-8
Molecular Formula C6H2BrCl3O2S
Molecular Weight (g/mol) 324.39
MDL Number MFCD03094651
InChI Key CKJIKXAPXLPSCL-UHFFFAOYSA-N
Synonym 4-bromo-2,6-dichlorobenzene-1-sulfonyl chloride, 4-bromo-2,6-dichlorobenzenesulphonyl chloride, 4-bromo-2,6-dichlorophenyl chlorosulfone, pubchem11744, acmc-1adw5, 4-bromo-2,6-dichlorobenzenesulfonylchloride, 4-bromo-2,6-dichlorobenzensulfonyl chloride, 4-bromo-2,6-dichlorophenylsulfonyl chloride, benzenesulfonylchloride, 4-bromo-2,6-dichloro, 4-bromanyl-2,6-bis chloranyl benzenesulfonyl chloride
PubChem CID 2737466
IUPAC Name 4-bromo-2,6-dichlorobenzenesulfonyl chloride
SMILES ClC1=CC(Br)=CC(Cl)=C1S(Cl)(=O)=O

Specifications

Melting Point 42°C to 48°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 324.41
Percent Purity 95%
Chemical Name or Material 4-Bromo-2,6-dichlorobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only