accessibility menu, dialog, popup

UserName

CAS RN 351003-54-8

IUPAC Name: 4-bromo-2,6-dichlorobenzene-1-sulfonyl chloride
Synonyms: 4-bromo-2,6-dichlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 324.39
Molecular Formula: C6H2BrCl3O2S
InChi Key: CKJIKXAPXLPSCL-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CC(Cl)=C1S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)