missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-difluoronitrobenzene 98.0+%, TCI America™

Catalog No. B39261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39261G 1 g
B39265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39261G Supplier TCI America Supplier No. B39261G

Chemical Identifiers

CAS 167415-27-2
Molecular Formula C6H2BrF2NO2
Molecular Weight (g/mol) 237.99
MDL Number MFCD00042184
InChI Key GJFYMYJYPARISZ-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluoronitrobenzene, 4-bromo-2,5-difluoro-1-nitrobenzene, benzene, 1-bromo-2,5-difluoro-4-nitro, buttpark 29\01-58, 1-bromo-2,5-difluoro-4-nitro-benzene, pubchem8498, zlchem 1046, acmc-209dwg, intermediates-zcf02256, ksc495e4h
PubChem CID 2736286
IUPAC Name 1-bromo-2,5-difluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(Br)C=C1F

Specifications

Melting Point 58°C
Color Yellow
Quantity 1 g
Formula Weight 237.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,5-difluoronitrobenzene