missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-difluoroaniline, 98%, Thermo Scientific Chemicals

$156.07 - $156.07

Chemical Identifiers

CAS 112279-60-4
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.006
MDL Number MFCD01861125
InChI Key XOYHFIQPPOJMFK-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluoro-phenylamine, 4-bromo-2,5-difluoro-aniline, 4-bromo-2,5-difluorophenylamine, pubchem2928, acmc-1bvn4, ksc494m7l, 4-bromo-2,5-difluorobenzenamine, 2,5-difluoro-4-bromoaniline, 4-bromo-2,5-difluoroaniline, benzenamine, 4-bromo-2,5-difluoro
PubChem CID 2773285
IUPAC Name 4-bromo-2,5-difluoroaniline
SMILES C1=C(C(=CC(=C1F)Br)F)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2445709
View Documents
Thermo Scientific Chemicals
B2445709
10 g
Each for $156.07
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112279-60-4
208.006
XOYHFIQPPOJMFK-UHFFFAOYSA-N
2773285
C1=C(C(=CC(=C1F)Br)F)N
C6H4BrF2N
MFCD01861125
4-bromo-2,5-difluoro-phenylamine, 4-bromo-2,5-difluoro-aniline, 4-bromo-2,5-difluorophenylamine, pubchem2928, acmc-1bvn4, ksc494m7l, 4-bromo-2,5-difluorobenzenamine, 2,5-difluoro-4-bromoaniline, 4-bromo-2,5-difluoroaniline, benzenamine, 4-bromo-2,5-difluoro
4-bromo-2,5-difluoroaniline

Specifications

112279-60-4
C6H4BrF2N
10 g
4-bromo-2,5-difluoro-phenylamine, 4-bromo-2,5-difluoro-aniline, 4-bromo-2,5-difluorophenylamine, pubchem2928, acmc-1bvn4, ksc494m7l, 4-bromo-2,5-difluorobenzenamine, 2,5-difluoro-4-bromoaniline, 4-bromo-2,5-difluoroaniline, benzenamine, 4-bromo-2,5-difluoro
C1=C(C(=CC(=C1F)Br)F)N
208.006
208.01
4-Bromo-2,5-difluoroaniline
74°C to 75°C
MFCD01861125
4741102
XOYHFIQPPOJMFK-UHFFFAOYSA-N
4-bromo-2,5-difluoroaniline
2773285
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only