missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,3-difluorophenol 98.0+%, TCI America™
$29.83 - $102.52
Chemical Identifiers
| CAS | 144292-32-0 |
|---|---|
| Molecular Formula | C6H3BrF2O |
| Molecular Weight (g/mol) | 208.99 |
| MDL Number | MFCD08061907 |
| InChI Key | JZAVCMMYGSROJP-UHFFFAOYSA-N |
| Synonym | 2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol |
| PubChem CID | 10219942 |
| IUPAC Name | 4-bromo-2,3-difluorophenol |
| SMILES | OC1=C(F)C(F)=C(Br)C=C1 |
Chemical Identifiers
| 144292-32-0 | |
| 208.99 | |
| JZAVCMMYGSROJP-UHFFFAOYSA-N | |
| 10219942 | |
| OC1=C(F)C(F)=C(Br)C=C1 |
| C6H3BrF2O | |
| MFCD08061907 | |
| 2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol | |
| 4-bromo-2,3-difluorophenol |
Specifications
| 144292-32-0 | |
| C6H3BrF2O | |
| 1 g | |
| JZAVCMMYGSROJP-UHFFFAOYSA-N | |
| 4-bromo-2,3-difluorophenol | |
| 10219942 | |
| ≥98.0% (GC,T) | |
| 4-Bromo-2,3-difluorophenol |
| 55°C | |
| MFCD08061907 | |
| 2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol | |
| OC1=C(F)C(F)=C(Br)C=C1 | |
| 208.99 | |
| 208.99 | |
| Crystalline Powder |
Safety and Handling
TSCA : No