missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,3-difluorophenol 98.0+%, TCI America™

Catalog No. D39131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39131G 1 g
D39135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39131G Supplier TCI America Supplier No. D39131G

Chemical Identifiers

CAS 144292-32-0
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD08061907
InChI Key JZAVCMMYGSROJP-UHFFFAOYSA-N
Synonym 2,3-difluoro-4-bromophenol, 4-bromo-2,3-difluoro-phenol, phenol, 4-bromo-2,3-difluoro, pubchem2477, pubchem4111, acmc-209csf, bromodifluorophenol, ksc494o1n, 2,3-difluoro 4-bromo phenol, 2.3-difluoro-4-bromo phenol
PubChem CID 10219942
IUPAC Name 4-bromo-2,3-difluorophenol
SMILES OC1=C(F)C(F)=C(Br)C=C1

Specifications

Melting Point 55°C
Quantity 1 g
Formula Weight 208.99
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,3-difluorophenol