missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,2-diphenylbutyronitrile, 95%

Catalog No. AAA1778622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1778622 100 g
AAA1778636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1778622 Supplier Thermo Scientific Chemicals Supplier No. A1778622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39186-58-8 | C16H14BrN | 300.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39186-58-8
Molecular Formula C16H14BrN
Molecular Weight (g/mol) 300.199
MDL Number MFCD00001845
InChI Key IGYSFJHVFHNOEI-UHFFFAOYSA-N
Synonym 4-bromo-2,2-diphenylbutyronitrile, 2,2-diphenyl-4-bromobutyronitrile, 4-bromo-2,2-diphenyl-butyronitrile, 3-cyano-3,3-diphenylpropyl bromide, benzeneacetonitrile, .alpha.-2-bromoethyl-.alpha.-phenyl, acmc-1csza, butyronitrile,2-diphenyl, ksc495i2r, aronis25014, bromoethyl diphenyl acetonitrile
PubChem CID 96575
IUPAC Name 4-bromo-2,2-diphenylbutanenitrile
SMILES C1=CC=C(C=C1)C(CCBr)(C#N)C2=CC=CC=C2

Specifications

Melting Point 67°C to 69°C
Quantity 100 g
UN Number UN3439
Beilstein 1684952
Formula Weight 300.2
Percent Purity 95%
Chemical Name or Material 4-Bromo-2,2-diphenylbutyronitrile
Hazard Category H302
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 254-337-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only