missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-imidazole 98.0+%, TCI America™

Catalog No. B32801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32801G 1 g
B32805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32801G Supplier TCI America Supplier No. B32801G

Chemical Identifiers

CAS 2302-25-2
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD00047021
InChI Key FHZALEJIENDROK-UHFFFAOYSA-N
Synonym 4-bromo-1h-imidazole, 4-bromoimidazole, 1h-imidazole, 4-bromo, 4 or 5-bromoimidazole, imidazole, 4-bromo, 4-bromo-3h-imidazole, imidazole, 4 or 5-bromo-6ci,7ci, 5-bromoimidazole, 3kqm, 4-bromo-imidazole
PubChem CID 96125
IUPAC Name 5-bromo-1H-imidazole
SMILES BrC1=CN=CN1

Specifications

Melting Point 133°C
Quantity 1 g
UN Number 2811
Formula Weight 146.98
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1H-imidazole
RTECSNumber NI3517450
TSCA No