missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-trityl-1H-pyrazole, 95%

Catalog No. p-7056970
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6622503 1 g
AAH6622506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6622503 Supplier Thermo Scientific Chemicals Supplier No. H6622503
Only null left

CAS: 95162-14-4 | C22H17BrN2 | 389.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95162-14-4
Molecular Formula C22H17BrN2
Molecular Weight (g/mol) 389.30
MDL Number MFCD09907863
InChI Key CPENTLJGGGSVAJ-UHFFFAOYSA-N
Synonym 4-bromo-1-trityl-1h-pyrazole, 1h-pyrazole, 4-bromo-1-triphenylmethyl, 1h-pyrazole,4-bromo-1-triphenylmethyl, 4-bromo-1-triphenylmethyl-1h-pyrazole, 4-bromo-1-triphenylmethyl pyrazole, n-trityl-4-bromopyrazole, 4-bromo-1-n-tritylpyrazole, 1-trityl-4-bromo-1h-pyrazole, 4-bromo-1-trityl-1-h-pyrazole
PubChem CID 10834188
IUPAC Name 4-bromo-1-(triphenylmethyl)-1H-pyrazole
SMILES BrC1=CN(N=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Beilstein 550183
Formula Weight 389.3
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Bromo-1-trityl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only