missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-5-carboxaldehyde, 97%

Catalog No. AAH3369003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3369003 1 g
AAH3369006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3369003 Supplier Thermo Scientific Chemicals Supplier No. H3369003

CAS: 473528-88-0 | C5H5BrN2O | 189.012 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 473528-88-0
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.012
MDL Number MFCD03074341
InChI Key GXAHYXQWHWDEDY-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-5-carbaldehyde, 4-bromo-1-methylpyrazole-5-carbaldehyde, 4-bromo-2-methyl-2h-pyrazole-3-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 4-bromo-1-methyl, acmc-1akfs, 4-bromo-1-methylpyrazole-5-carboxaldehyde, 4-bromo-1-methyl-1h-pyrazole-5-carboxaldehyde, 4-bromo-2-methyl-2 h-pyrazole-3-carbaldehyde
PubChem CID 2794634
IUPAC Name 4-bromo-2-methylpyrazole-3-carbaldehyde
SMILES CN1C(=C(C=N1)Br)C=O

Specifications

Melting Point 97°C to 98°C
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 189.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-5-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only