accessibility menu, dialog, popup

UserName

CAS RN 473528-88-0

IUPAC Name: 4-bromo-1-methyl-1H-pyrazole-5-carbaldehyde
Synonyms: 4-bromo-2-methylpyrazole-3-carbaldehyde
Molecular Weight (g/mol): 189.01
Molecular Formula: C5H5BrN2O
InChi Key: GXAHYXQWHWDEDY-UHFFFAOYSA-N
SMILES: CN1N=CC(Br)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results