missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-4391216
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC63901CB 250mg
CC63901DA 1g
CC63901DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC63901CB Supplier Thermo Scientific Chemicals Supplier No. CC63901CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84547-86-4
Molecular Formula C5H5BrN2O2
Molecular Weight (g/mol) 205.011
InChI Key LEEPGDCCHVRYHK-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-bromo-1-methyl, 4-bromo-1-methyl-1h-pyrazole-3-carboxylicacid, pubchem16814, 4-bromo-1-methyl-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 4-bromo-1-methyl, 4-bromo-1-methyl-1h-pyrazole-3-carboxylic acid #
PubChem CID 536013
IUPAC Name 4-bromo-1-methylpyrazole-3-carboxylic acid
SMILES CN1C=C(C(=N1)C(=O)O)Br

Specifications

Assay Percent Range 97%
Quantity 250mg
Formula Weight 205.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid
Hazard Category GHS Signal Word: Warning