missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific Chemicals

Catalog No. AAH3313103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3313103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3313103 Supplier Thermo Scientific Chemicals Supplier No. H3313103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 287922-71-8 | Molecular Formula: C5H4BrN3 | Molecular Weight: 186.01 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 287922-71-8
Molecular Formula C5H4BrN3
Molecular Weight (g/mol) 186.01
MDL Number MFCD00103545
InChI Key NTTLCOOFUGHMJS-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carbonitrile, 1h-pyrazole-3-carbonitrile, 4-bromo-1-methyl, 4-bromo-1-methyl-pyrazole-3-carbonitrile
PubChem CID 45594322
IUPAC Name 4-bromo-1-methylpyrazole-3-carbonitrile
SMILES CN1C=C(Br)C(=N1)C#N

Specifications

Melting Point 103°C to 105°C
Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 186.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only