CAS RN 287922-71-8
IUPAC Name:
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile
Synonyms:
4-bromo-1-methylpyrazole-3-carbonitrile
Molecular Weight (g/mol):
186.01
Molecular Formula:
C5H4BrN3
InChi Key:
NTTLCOOFUGHMJS-UHFFFAOYSA-N
SMILES:
CN1C=C(Br)C(=N1)C#N
1
–
1
of
1
results
1
Promotions Available
4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific Chemicals
4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific Chemicals
CAS: 287922-71-8 Molecular Formula: C5H4BrN3 Molecular Weight (g/mol): 186.01 MDL Number: MFCD00103545 InChI Key: NTTLCOOFUGHMJS-UHFFFAOYSA-N Synonym: 4-bromo-1-methyl-1h-pyrazole-3-carbonitrile,1h-pyrazole-3-carbonitrile, 4-bromo-1-methyl,4-bromo-1-methyl-pyrazole-3-carbonitrile PubChem CID: 45594322 IUPAC Name: 4-bromo-1-methylpyrazole-3-carbonitrile SMILES: CN1C=C(Br)C(=N1)C#N