missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene 98.0+%, TCI America™

Catalog No. p-7123604
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45391G 1 g
B45395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45391G Supplier TCI America Supplier No. B45391G
Only null left

Chemical Identifiers

CAS 173336-76-0
Molecular Formula C11H15BrO3
Molecular Weight (g/mol) 275.142
MDL Number MFCD14582816
InChI Key JFSCCLJKKCRXSS-UHFFFAOYSA-N
PubChem CID 21458711
IUPAC Name 4-bromo-1-methoxy-2-(3-methoxypropoxy)benzene
SMILES COCCCOC1=C(C=CC(=C1)Br)OC

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 129°C
Quantity 1 g
Formula Weight 275.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene
TSCA No
Recommended Storage Refrigerator