missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1-indanone 97.0+%, TCI America™
$44.28 - $82.79
Chemical Identifiers
| CAS | 15115-60-3 |
|---|---|
| Molecular Formula | C9H7BrO |
| Molecular Weight (g/mol) | 211.06 |
| MDL Number | MFCD01719772 |
| InChI Key | UVVYFYLSZIMKMC-UHFFFAOYSA-N |
| Synonym | 4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone |
| PubChem CID | 98713 |
| IUPAC Name | 4-bromo-2,3-dihydro-1H-inden-1-one |
| SMILES | BrC1=CC=CC2=C1CCC2=O |
Chemical Identifiers
| 15115-60-3 | |
| 211.06 | |
| UVVYFYLSZIMKMC-UHFFFAOYSA-N | |
| 98713 | |
| BrC1=CC=CC2=C1CCC2=O |
| C9H7BrO | |
| MFCD01719772 | |
| 4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone | |
| 4-bromo-2,3-dihydro-1H-inden-1-one |
Specifications
| 15115-60-3 | |
| 125°C | |
| MFCD01719772 | |
| 4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone | |
| BrC1=CC=CC2=C1CCC2=O | |
| 211.06 | |
| 211.06 | |
| Crystalline Powder |
| 94°C | |
| C9H7BrO | |
| 1 g | |
| UVVYFYLSZIMKMC-UHFFFAOYSA-N | |
| 4-bromo-2,3-dihydro-1H-inden-1-one | |
| 98713 | |
| ≥97.0% (GC) | |
| 4-Bromo-1-indanone |
Safety and Handling
RTECSNumber : NK7536100
TSCA : No