missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-fluoro-2-nitrobenzene 96.0+%, TCI America™

Catalog No. p-7119464
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
B20751G 1 g
B20755G 5 g
B207525G 25 g
3 options

Catalog No. B20751G

Supplier: TCI America B20751G

Only null left

Chemical Identifiers

CAS 364-73-8
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD00129165
InChI Key UQEANKGXXSENNF-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoronitrobenzene, 1-bromo-4-fluoro-3-nitrobenzene, 2-fluoro-5-bromonitrobenzene, 4-bromo-1-fluoro-2-nitro-benzene, benzene, 4-bromo-1-fluoro-2-nitro, pubchem1046, 5-bromo-fluoronitrobenzene, acmc-1cpc6, 5-bromo-2-fluronitrobenzene, ksc226i7f
PubChem CID 2736328
IUPAC Name 4-bromo-1-fluoro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Br)=CC=C1F

Specifications

Melting Point 19°C
Boiling Point 147°C
Quantity 1 g
Formula Weight 220.00
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-fluoro-2-nitrobenzene