missing translation for 'onlineSavingsMsg'
Learn More

4-bromo-1-fluoro-2-nitrobenzene, 98%

Catalog No. p-3212006
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC380890050 5 g Glass bottle
AC380890250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC380890050 Supplier Thermo Scientific Chemicals Supplier No. 380890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 364-73-8 | C6H3BrFNO2 | 220.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 364-73-8
Molecular Formula C6H3BrFNO2
Molecular Weight (g/mol) 220.00
MDL Number MFCD00129165
InChI Key UQEANKGXXSENNF-UHFFFAOYSA-N
Synonym 5-bromo-2-fluoronitrobenzene, 1-bromo-4-fluoro-3-nitrobenzene, 2-fluoro-5-bromonitrobenzene, 4-bromo-1-fluoro-2-nitro-benzene, benzene, 4-bromo-1-fluoro-2-nitro, pubchem1046, 5-bromo-fluoronitrobenzene, acmc-1cpc6, 5-bromo-2-fluronitrobenzene, ksc226i7f
PubChem CID 2736328
IUPAC Name 4-bromo-1-fluoro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Br)=CC=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 18°C to 20°C
Color Brown to Yellow
Density 1.7860g/mL
Boiling Point 240°C to 241°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5750 to 1.578
Linear Formula BrC6H3(F)NO2
Quantity 5 g
Specific Gravity 1.786
Formula Weight 220
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 4-Bromo-1-fluoro-2-nitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only